1,4-Benzodioxane-6-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,4-Benzodioxane-6-boronic acid, pinacol ester, 2,3-Dihydro-1,4-benzodioxine-6-boronic acid, pinacol ester, 2-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

11097-25G 11097-5G
76882-110EA 587.74 USD
76882-110 76882-112
1,4-Benzodioxane-6-boronic acid pinacol ester ≥97%
1,4-Benzodioxane-6-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₄
MW: 262.1093 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663890
CAS Number: 517874-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR