2,3-Difluoro-4-hexyloxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Difluoro-4-(n-hexyloxy)phenylboronic acid

10548-25G 10548-5G
76882-058EA 524.09 USD
76882-058 76882-060
2,3-Difluoro-4-hexyloxyphenylboronic acid ≥97%
2,3-Difluoro-4-hexyloxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₇BF₂O₃
MW: 258.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06656273
CAS Number: 121219-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR