1-(2,4-Difluoro-5-hydroxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
101985-1G
101985-500MG
101985-5G
76882-032EA
792.5
USD
76882-032
76882-034
76882-036
1-(2,4-Difluoro-5-hydroxyphenyl)ethanone ≥97%
1-(2,4-Difluoro-5-hydroxyphenyl)ethanone
Formula:
C₈H₆F₂O₂ MW: 172.129 g/mol Storage Temperature: Ambient |
CAS Number:
1807179-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...