2,4-Dibenzyloxy-5-pyrimidineboronic acid 97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Bis(benzyloxy)pyrimidine-5-boronic acid, 2,4-Bis(benzyloxy)pyrimidin-5-ylboronic acid

10163-1G 10163-25G 10163-5G 10163-500MG
76882-012EA 79.57 USD
76882-012 76882-014 76882-016 77644-276
2,4-Dibenzyloxy-5-pyrimidineboronic acid 97%
2,4-Dibenzyloxy-5-pyrimidineboronic acid
Formula: C₁₈H₁₇BN₂O₄
MW: 336.150 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318938
CAS Number: 70523-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR