(6-(Methylcarbamoyl)pyridin-3-yl)boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

10029-1G 10029-25G 10029-500MG 10029-5G
76882-002EA 231.28 USD
76882-002 76882-004 76882-006 76882-008
(6-(Methylcarbamoyl)pyridin-3-yl)boronic acid ≥95% research grade
(6-(Methylcarbamoyl)pyridin-3-yl)boronic acid
Formula: C₇H₉BN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08458474
CAS Number: 1217340-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR