2-Ethoxypyrimidine-5-carboxylic acid

Supplier: AOBChem USA

Warning

62522-1G 62522-25G 62522-500MG 62522-5G
76905-558EA 257.8 USD
76905-558 76905-560 76905-562 76905-564
2-Ethoxypyrimidine-5-carboxylic acid
2-Ethoxypyrimidine-5-carboxylic acid
Formula: C₇H₈N₂O₃
Storage Temperature: Ambient
CAS Number: 1242458-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR