4-Bromo-2,3-dimethylbenzaldehyde

Supplier: AOBChem USA

Warning

14048-1G 14048-25G 14048-500MG 14048-5G
76907-356EA 201.58 USD
76907-356 76907-358 76907-360 76907-362
4-Bromo-2,3-dimethylbenzaldehyde
4-Bromo-2,3-dimethylbenzaldehyde
Formula: C₉H₉BrO
MW: 213.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835096
CAS Number: 1114808-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR