2-(Methylthio)-5-pyrimidineboronic acid research grade

Supplier: AOBChem USA

Warning

Synonyms: (2-(Methylthio)pyrimidin-5-yl)boronic acid, 2-(Methylthio)pyrimidine-5-boronic acid

38835-1G 38835-25G 38835-5G
76889-746EA 64.72 USD
76889-746 76889-748 76889-750
2-(Methylthio)-5-pyrimidineboronic acid research grade
2-(Methylthio)-5-pyrimidineboronic acid
Formula: C₅H₇BN₂O₂S
MW: 170.0000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375142
CAS Number: 348098-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR