Ethyl 2-fluoropyridine-3-carboxylate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-fluoronicotinate

62470-1G 62470-25G 62470-5G
76943-680EA 100.79 USD
76943-680 76943-682 76943-684
Ethyl 2-fluoropyridine-3-carboxylate ≥97% research grade
Ethyl 2-fluoropyridine-3-carboxylate
Formula: C₈H₈FNO₂
MW: 169.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698241
CAS Number: 113898-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR