Ethyl 3-hydroxy-3-methylbutanoate ≥97%

Supplier: AOBChem USA

Warning

55553-1G 55553-25G 55553-5G
76943-236EA 94.43 USD
76943-236 76943-238 76943-240
Ethyl 3-hydroxy-3-methylbutanoate ≥97%
Ethyl 3-hydroxy-3-methylbutanoate
Formula: C₇H₁₄O₃
MW: 146.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16749012
CAS Number: 18267-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR