Ethyl 2-(1-hydroxycyclobutyl)acetate ≥97% research grade

Supplier: AOBChem USA

Warning

55395-1G 55395-25G 55395-500MG 55395-5G
76943-220EA 177.18 USD
76943-220 76943-222 76943-224 76943-226
Ethyl 2-(1-hydroxycyclobutyl)acetate ≥97% research grade
Ethyl 2-(1-hydroxycyclobutyl)acetate
Formula: C₈H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD16749053
CAS Number: 27784-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR