6-Chloro-3-iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

54793-1G 54793-25G 54793-500MG 54793-5G
76943-180EA 255.68 USD
76943-180 76943-182 76943-184 76943-186
6-Chloro-3-iodo-2-methoxypyridine ≥97%
6-Chloro-3-iodo-2-methoxypyridine
Formula: C₆H₅ClINO
Storage Temperature: Ambient
MDL Number: MFCD18415726
CAS Number: 1261769-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR