Methyl 3-formyl-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

45653-1G 45653-25G 45653-5G
76942-442EA 117.76 USD
76942-442 76942-444 76942-446
Methyl 3-formyl-4-methylbenzoate ≥97%
Methyl 3-formyl-4-methylbenzoate
Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24678288
CAS Number: 23038-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR