6-Ethoxypyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

37190-1G 37190-25G 37190-500MG 37190-5G
76941-730EA 346.92 USD
76941-730 76941-732 76941-734 76941-736
6-Ethoxypyridin-3-ol ≥97%
6-Ethoxypyridin-3-ol

Precaución: for Research Use Only

Formula: C₇H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18819177
CAS Number: 116178-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR