Methyl 2-bromo-6-fluoro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

32212-1G 32212-25G 32212-5G
76940-124EA 94.43 USD
76940-124 76940-126 76940-128
Methyl 2-bromo-6-fluoro-4-methylbenzoate ≥97%
Methyl 2-bromo-6-fluoro-4-methylbenzoate
Formula: C₉H₈BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28579975
CAS Number: 1807232-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR