6-Phenylpicolinaldehyde ≥97%

Supplier: AOBChem USA

Warning

25288-1G 25288-25G 25288-500MG 25288-5G
76938-582EA 126.25 USD
76938-582 76938-584 76938-586 76938-588
6-Phenylpicolinaldehyde ≥97%
6-Phenylpicolinaldehyde
Formula: C₁₂H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD11044383
CAS Number: 157402-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR