4-Iodo-1,3,5-trimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

57245-25G 57245-5G
76934-356EA 559.1 USD
76934-356 76934-358
4-Iodo-1,3,5-trimethyl-1H-pyrazole ≥97%
4-Iodo-1,3,5-trimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₉IN₂
Storage Temperature: Ambient
MDL Number: MFCD00626320
CAS Number: 51660-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR