5-Bromo-1-chloro-2-fluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

48935-1G 48935-25G 48935-500MG 48935-5G
76932-848EA 439.22 USD
76932-848 76932-850 76932-852 76932-854
5-Bromo-1-chloro-2-fluoro-3-methoxybenzene ≥97%
5-Bromo-1-chloro-2-fluoro-3-methoxybenzene
Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31555542
CAS Number: 1784628-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR