4-Fluoro-3-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46975-1G 46975-25G 46975-5G
76932-698EA 73.21 USD
76932-698 76932-700 76932-702
4-Fluoro-3-iodobenzaldehyde ≥97%
4-Fluoro-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄OFI
MW: 250.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236850
CAS Number: 227609-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR