4-Ethoxy-2,3-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

37445-25G 37445-5G
76932-052EA 346.92 USD
76932-052 76932-054
4-Ethoxy-2,3-difluorophenol ≥97%
4-Ethoxy-2,3-difluorophenol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07368025
CAS Number: 126163-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR