4-Bromo-2-fluoro-6-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

33131-1G 33131-25G 33131-500MG 33131-5G
76931-116EA 334.19 USD
76931-116 76931-118 76931-120 76931-122
4-Bromo-2-fluoro-6-methoxyphenol ≥97%
4-Bromo-2-fluoro-6-methoxyphenol
Formula: C₇H₆BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28641990
CAS Number: 1781791-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR