4-Bromo-3-chloro-2-methylphenol ≥97% research grade

Supplier: AOBChem USA

Warning

30023-1G 30023-25G 30023-500MG 30023-5G
76930-136EA 260.99 USD
76930-136 76930-138 76930-140 76930-142
4-Bromo-3-chloro-2-methylphenol ≥97% research grade
4-Bromo-3-chloro-2-methylphenol
Formula: C₇H₆BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD22414510
CAS Number: 55289-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR