4-Bromo-2-phenylaniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-[1,1'-biphenyl]-2-amine

27600-1G 27600-25G 27600-500MG 27600-5G
76929-586EA 257.8 USD
76929-586 76929-588 76929-590 76929-592
4-Bromo-2-phenylaniline ≥97%
4-Bromo-2-phenylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrN
MW: 248.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09754386
CAS Number: 5455-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR