4-Ethoxy-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

25534-1G 25534-25G 25534-500MG 25534-5G
76928-812EA 221.73 USD
76928-812 76928-814 76928-816 76928-818
4-Ethoxy-2-fluoropyridine ≥97%
4-Ethoxy-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₇H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD09835274
CAS Number: 175965-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR