4-Chloro-3,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18803-1G 18803-25G 18803-500MG 18803-5G
76926-572EA 142.17 USD
76926-572 76926-574 76926-576 76926-578
4-Chloro-3,5-dimethylbenzaldehyde ≥97%
4-Chloro-3,5-dimethylbenzaldehyde
Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111043
CAS Number: 51719-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR