5-Bromo-3-iodo-2-isopropoxypyridine ≥97%

Supplier: AOBChem USA

Warning

17419-1G 17419-25G 17419-500MG 17419-5G
76926-186EA 127.31 USD
76926-186 76926-188 76926-190 76926-192
5-Bromo-3-iodo-2-isopropoxypyridine ≥97%
5-Bromo-3-iodo-2-isopropoxypyridine
Formula: C₈H₉BrINO
MW: 341.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07366719
CAS Number: 848243-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR