5-Bromo-2-methoxypyridin-4-ol ≥97%

Supplier: AOBChem USA

Warning

115014-1G 115014-25G 115014-500MG 115014-5G
76924-590EA 341.61 USD
76924-590 76924-592 76924-594 76924-596
5-Bromo-2-methoxypyridin-4-ol ≥97%
5-Bromo-2-methoxypyridin-4-ol
Formula: C₆H₆BrNO₂
MW: 204.021 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22684720
CAS Number: 1420034-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR