2-Bromo-3-chlorophenol ≥97%

Supplier: AOBChem USA

Warning

63274-25G 63274-5G
76905-612EA 410.57 USD
76905-612 76905-614
2-Bromo-3-chlorophenol ≥97%
2-Bromo-3-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrClO
MW: 207.452 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037216
CAS Number: 855836-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR