2-Isopropoxybenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
56021-1G
56021-25G
56021-5G
76905-176EA
62.6
USD
76905-176
76905-178
76905-180
2-Isopropoxybenzaldehyde ≥97%
2-Isopropoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01571834 CAS Number: 22921-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...