2-Bromo-1-(difluoromethoxy)-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

53228-1G 53228-25G 53228-5G
76904-910EA 96.55 USD
76904-910 76904-912 76904-914
2-Bromo-1-(difluoromethoxy)-3-fluorobenzene ≥97%
2-Bromo-1-(difluoromethoxy)-3-fluorobenzene
Formula: C₇H₄BrF₃O
MW: 241.005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18390895
CAS Number: 1239492-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR