2-Fluoro-5-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

49353-25G 49353-5G
76904-466EA 524.09 USD
76904-466 76904-468
2-Fluoro-5-isopropoxybenzoic acid ≥97%
2-Fluoro-5-isopropoxybenzoic acid
Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356760
CAS Number: 219939-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR