2-Biphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-([1,1'-Biphenyl]-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

35688-25G 35688-5G
76903-184EA 346.92 USD
76903-184 76903-186
2-Biphenylboronic acid pinacol ester ≥97%
2-Biphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₁BO₂
MW: 280.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14636459
CAS Number: 914675-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR