2,6-Difluoro-3-formylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

33752-1G 33752-25G 33752-500MG 33752-5G
76902-712EA 338.43 USD
76902-712 76902-714 76902-716 76902-718
2,6-Difluoro-3-formylbenzonitrile ≥97%
2,6-Difluoro-3-formylbenzonitrile
Formula: C₈H₃F₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD26687933
CAS Number: 1541976-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR