2-Formyl-5-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33704-1G 33704-25G 33704-500MG 33704-5G
76902-668EA 124.13 USD
76902-668 76902-670 76902-672 76902-674
2-Formyl-5-methylphenylboronic acid pinacol ester ≥97%
2-Formyl-5-methylphenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₃
MW: 246.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18731005
CAS Number: 1422268-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR