2-Methoxy-4-(trifluoromethyl)benzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
29204-25G
29204-5G
76901-160EA
381.93
USD
76901-160
76901-162
2-Methoxy-4-(trifluoromethyl)benzaldehyde ≥97%
2-Methoxy-4-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₉H₇F₃O₂ MW: 204.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115975 CAS Number: 132927-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...