2-Bromo-N,N-dimethylbenzylamine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2-bromophenyl)-N,N-dimethylmethanamine, (2-Bromophenyl)-N,N-dimethylmethylamine

27205-1G 27205-25G 27205-5G
76900-632EA 62.6 USD
76900-632 76900-634 76900-636
2-Bromo-N,N-dimethylbenzylamine ≥97%
2-Bromo-N,N-dimethylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MW: 214.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010352
CAS Number: 1976-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR