2-Bromo-5-chloro-4-(hydroxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-5-chloro-4(hydroxymethyl)pyridine

24483-1G 24483-25G 24483-500MG 24483-5G
76899-484EA 260.99 USD
76899-484 76899-486 76899-488 76899-490
2-Bromo-5-chloro-4-(hydroxymethyl)pyridine ≥97%
2-Bromo-5-chloro-4-(hydroxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.467 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16606985
CAS Number: 1227599-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR