2-Chloro-5-hydroxy-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-4-methylpyridin-3-ol, 2-Chloro-5-hydroxy-4-picoline

20554-1G 20554-25G 20554-500MG 20554-5G
76898-038EA 201.58 USD
76898-038 76898-040 76898-042 76898-044
2-Chloro-5-hydroxy-4-methylpyridine ≥97%
2-Chloro-5-hydroxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.571 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610471
CAS Number: 1227502-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR