2-[4-(Trifluoromethoxy)phenyl]-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

15533-1G 15533-25G 15533-5G
76896-050EA 62.6 USD
76896-050 76896-052 76896-054
2-[4-(Trifluoromethoxy)phenyl]-1,3-dioxolane ≥97%
2-[4-(Trifluoromethoxy)phenyl]-1,3-dioxolane
Formula: C₁₀H₉F₃O₃
MW: 234.172 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209128
CAS Number: 773102-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR