2-Chloro-6-(methylsulfanyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

10953-1G 10953-25G 10953-500MG 10953-5G
76894-472EA 153.84 USD
76894-472 76894-474 76894-476 76894-478
2-Chloro-6-(methylsulfanyl)pyridine ≥97%
2-Chloro-6-(methylsulfanyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₆ClNS
Storage Temperature: Ambient
MDL Number: MFCD00173422
CAS Number: 77145-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR