4-Bromo-1-isobutyl-3,5-dimethyl-1H-pyrazole ≥97% research grade

Supplier: AOBChem USA

Warning

57255-1G 57255-25G 57255-500MG 57255-5G
76917-322EA 245.07 USD
76917-322 76917-324 76917-326 76917-328
4-Bromo-1-isobutyl-3,5-dimethyl-1H-pyrazole ≥97% research grade
4-Bromo-1-isobutyl-3,5-dimethyl-1H-pyrazole
Formula: C₉H₁₅BrN₂
MW: 231.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12027152
CAS Number: 1171526-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR