3-Cyano-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52728-1G 52728-25G 52728-500MG 52728-5G
76916-898EA 135.8 USD
76916-898 76916-900 76916-902 76916-904
3-Cyano-4-methylbenzoic acid ≥97%
3-Cyano-4-methylbenzoic acid
Formula: C₉H₇NO₂
MW: 161.157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00045859
CAS Number: 138642-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR