3-Bromo-5-(4-fluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

26944-1G 26944-25G 26944-500MG 26944-5G
76912-592EA 141.1 USD
76912-592 76912-594 76912-596 76912-598
3-Bromo-5-(4-fluorophenyl)pyridine ≥97%
3-Bromo-5-(4-fluorophenyl)pyridine
Formula: C11H7BrFN
Storage Temperature: Ambient
MDL Number: MFCD09800589
CAS Number: 675590-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR