3-Bromo-5-(1-methylethyl)-benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

24781-1G 24781-25G 24781-500MG 24781-5G
76911-816EA 201.58 USD
76911-816 76911-818 76911-820 76911-822
3-Bromo-5-(1-methylethyl)-benzenemethanol ≥97%
3-Bromo-5-(1-methylethyl)-benzenemethanol
Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28977156
CAS Number: 1781019-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR