4-Amino-3,5-difluorobenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

24064-1G 24064-25G 24064-500MG 24064-5G
76911-478EA 334.19 USD
76911-478 76911-480 76911-482 76911-484
4-Amino-3,5-difluorobenzeneboronic acid pinacol ester ≥97%
4-Amino-3,5-difluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BF₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18434474
CAS Number: 939968-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR