3-Ethoxy-2,6-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21519-25G 21519-5G
76910-528EA 354.35 USD
76910-528 76910-530
3-Ethoxy-2,6-difluorobenzaldehyde ≥97%
3-Ethoxy-2,6-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11519366
CAS Number: 1028263-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR