4-Bromo-2-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16506-1G 16506-25G 16506-500MG 16506-5G
76908-540EA 270.53 USD
76908-540 76908-542 76908-544 76908-546
4-Bromo-2-(trifluoromethoxy)phenylboronic acid ≥97%
4-Bromo-2-(trifluoromethoxy)phenylboronic acid
Formula: C₇H₅BBrF₃O₃
MW: 284.823 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441792
CAS Number: 1048990-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR