3-Chloro-2-methylbenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Chloro-2-methyl-3-(trifluoromethyl)benzene

62362-1G 62362-25G 62362-5G
76892-582EA 79.57 USD
76892-582 76892-584 76892-586
3-Chloro-2-methylbenzotrifluoride ≥97%
3-Chloro-2-methylbenzotrifluoride
Formula: C₈H₆ClF₃
MW: 194.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226662
CAS Number: 93242-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR