(2,5-Dichloro-4-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

61660-1G 61660-500MG 61660-5G 61660-25G
76892-512EA 461.5 USD
76892-512 76892-514 76892-516 77656-700
(2,5-Dichloro-4-fluorophenyl)boronic acid ≥97%
(2,5-Dichloro-4-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₄BCl₂FO₂
MW: 208.8101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786329
CAS Number: 3028870-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR