1-Isopropyl-1H-pyrazole-5-carboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-Isopropylpyrazole-5-carboxylic Acid
56554-1G
56554-25G
56554-500MG
56554-5G
76891-908EA
126.25
USD
76891-908
76891-910
76891-912
76891-914
1-Isopropyl-1H-pyrazole-5-carboxylic acid ≥97%
1-Isopropyl-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂O₂ MW: 154.167 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03130043 CAS Number: 920006-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...